missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Citrinin, MP Biomedicals™
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Specifications
Specifications
Color | Yellow |
Quantity | 1 mg |
Formula Weight | 250.3 |
Physical Form | Crystalline Powder |
Chemical Name or Material | Citrinin |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'