Learn More
DWK Life Sciences Clamp for Spherical Joints, with tightening screw, made of stainless steel
Ball joint clamp with additional locking option.
9.86 EUR - 708.00 EUR
Specifications
Material | Stainless Steel |
---|---|
For Use With (Equipment) | Securing of Spherical Ground Joints |
For Use With (Application) | Securing of spherical ground joints |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
10623982
|
DWK Life Sciences
286334202 |
S 35 | N/A | ||||||
15854678
|
DWK Life Sciences
286334802 |
S 51 |
489.00 EUR
Pack of 10 |
Estimated Shipment: 16-01-2025
Log in to see stock. |
|||||
15844678
|
DWK Life Sciences
286334605 |
S 40 and S 41 |
682.00 EUR
Pack of 25 |
Estimated Shipment: 16-01-2025
Log in to see stock. |
|||||
10409342
|
DWK Life Sciences
286333806 |
S 29 | N/A | ||||||
10141781
|
DWK Life Sciences
286333403 |
S 19 |
9.86 EUR
Each |
Estimated Shipment: 17-01-2025
Log in to see stock. |
|||||
15864678
|
DWK Life Sciences
286335001 |
S 64 |
320.00 EUR
Pack of 5 |
Estimated Shipment: 16-01-2025
Log in to see stock. |
|||||
15921571
|
DWK Life Sciences
286333009 |
S 13 |
708.00 EUR
Pack of 50 |
Estimated Shipment: 15-01-2025
Log in to see stock. |
|||||
Description
The ball joint clamp with locking screw enables a very secure connection by additional locking with the locking screw.
- The locking screw enables a very secure connection by means of an additional locking device.
- Excellent dimensional stability compared to conventional wire clamps
Specifications
Stainless Steel | |
Securing of Spherical Ground Joints | |
Securing of spherical ground joints |