missing translation for 'onlineSavingsMsg'
Learn More

RETSCH Clamping Covers

158.00 EUR - 7485.00 EUR

Specifications

Packaging Quantity 1 Ea.
View More Specs

Products 3
Product Code Brand Product Type For Use With (Equipment) Price Quantity & Availability  
Product Code Brand Product Type For Use With (Equipment) Price Quantity & Availability  
10567601
View Documents
RETSCH
03.660.0006
Clamping lid without window Test sieves 100/150/200/203 mm dia. (only for use with threaded rods) This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10501075
View Documents
RETSCH
02.660.0012
Clamping lid with large window of Perspex Test Sieves 400 mm dia.
515.00 EUR
Each
Estimated Shipment: 24-02-2025
to see stock.
Add to basket
 
10354153
View Documents
RETSCH
02.660.0011
Clamping lid with large window of Perspex Test Sieves 305 mm dia.
7485.00 EUR
Each
Estimated Shipment: 24-02-2025
to see stock.
Add to basket
 
Description

Description

Specifications

Specifications

1 Ea.
Videos
SDS
Documents

Documents

Product Certifications