missing translation for 'onlineSavingsMsg'
Learn More

VITLAB™ Class A PMP Graduated Cylinders with Raised Scale

Brand:  VITLAB™ 64704

 View more versions of this product

Product Code. 11338543

  • 17.40 EUR / Pack of 2
Estimated Shipment: 13-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

A
±0.25mL
Hexagonal
Graduated Cylinder
0.5 mL
Round
170 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers