Learn More
Hirschmann™ Class AS Volumetric Pipettes with Amber Stain Graduation
Product Code. p-8013420
Click to view available options
Volume (Metric):
0.5 mL
1 mL
1.5 mL
2 mL
2.5 mL
3 mL
4 mL
5 mL
6 mL
7 mL
8 mL
9 mL
10 mL
15 mL
20 mL
25 mL
30 mL
40 mL
50 mL
100 mL
Unit Size:
Pack of 12
Pack of 6
Product Code. 11782252
Brand: Hirschmann™ 1340129
Only null left
Add to basket
Add to basket
Provides an extremely high degree of volumetric accuracy for your pipetting needs. Hirschmann Instruments™ Class AS Volumetric Pipettes with Amber Stain Graduation are manufactured from clear AR™ glass.
  • One mark
  • Acc. to DIN EN ISO 648
  • Conformity certified, with dated batch identification
Specifications
Class AS
Material AR-GLAS
Graduated Yes
Graduation Color Brown
Quantity 12/Pk.
Tolerance 0.005 mL
Volume (Metric) 0.5 mL
Length (Metric) 300 mm
Type Volumetric Pipet