Learn More
VITLAB™ Class B Polypropylene Short Form Measuring Cylinders
Use these cylinders for food industry and volume measurement applications. VITLAB™ Class B Polypropylene Short Form Measuring Cylinders feature easy to read, raised scale and ring marks at the primary scale points.
Brand: VITLAB™ 640941
This item is not returnable.
View return policy
Description
- Highly transparent polypropylene graduated cylinder
- Calibrated (In)
- Class B tolerances according to DIN 12681 / ISO 6706.
- Thermal stress up to 80°C does not cause tolerance limits to be permanently exceeded
Specifications
B | |
±0.5mL | |
122 mm |
0.5 mL | |
Round | |
Graduated Cylinder |