Learn More
Zwiebel™ Class F1 Weight Box with Certificate
173.00 EUR - 315.00 EUR
Specifications
For Use With (Application) For use with Weighing Instruments of Special Accuracy Class I and High Accuracy Class II
View More Specs
Products 3
Product Code Brand Mass Tolerance Price Quantity & Availability  
Product Code Brand Mass Tolerance Price Quantity & Availability  
11870892
View Documents
Zwiebel™
PBI56819
100 g ±5 mg
219.00 EUR
Each
Estimated Shipment: 21-02-2025
to see stock.
Add to basket
11880902
View Documents
Zwiebel™
PBI56822
2000 g ±10 mg
315.00 EUR
Each
Estimated Shipment: 21-02-2025
to see stock.
Add to basket
11810892
View Documents
Zwiebel™
PBI56812
500 g ±2.50 mg
173.00 EUR
Each
Estimated Shipment: 21-02-2025
to see stock.
Specifications
For use with Weighing Instruments of Special Accuracy Class I and High Accuracy Class II