Learn More
Zwiebel™ Class M1 Weight Set
Use with weighing instruments of medium accuracy class III. Zwiebel™ Class M1 Weights are ideal for the verification or calibration of class M2 weights.
74.68 EUR - 571.34 EUR
Specifications
For Use With (Application) | For use with Weighing Instruments of Medium Accuracy Class III and Ordinary Accuracy Class IIII |
---|
Product Code | Brand | Mass | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Mass | Price | Quantity & Availability | |||||
11830962
|
Zwiebel™
PBI56845 |
1 to 500 g |
240.62 EUR
Each |
Estimated Shipment: 05-11-2024
Log in to see stock. |
|||||
11850952
|
Zwiebel™
PBI5683W |
1 to 200 g |
199.28 EUR
Each |
Estimated Shipment: 29-11-2024
Log in to see stock. |
|||||
11810982
|
Zwiebel™
PBI5684Y |
1 to 2000 g |
571.34 EUR
Each |
Estimated Shipment: 22-11-2024
Log in to see stock. |
|||||
11880922
|
Zwiebel™
PBI5682T |
1 to 500 mg |
74.68 EUR
Each |
Estimated Shipment: 29-11-2024
Log in to see stock. |
|||||
11860932
|
Zwiebel™
PBI56835 |
1 to 50 g |
86.23 EUR
Each |
Estimated Shipment: 29-11-2024
Log in to see stock. |
|||||
Description
Specifications
For use with Weighing Instruments of Medium Accuracy Class III and Ordinary Accuracy Class IIII |