missing translation for 'onlineSavingsMsg'
Learn More

Zwiebel™ Class M1 Weight Set with Certificate

Use with weighing instruments of medium accuracy class III. Zwiebel™ Class M1 Weights are ideal for the verification or calibration of class M2 weights.

Brand:  Zwiebel™ PBI56836

 View more versions of this product

Product Code. 11870932

  • 146.00 EUR / Each
Estimated Shipment: 23-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Class M1 stainless steel calibration weight
  • Delivered with a calibration certificate guaranteeing traceability
  • In compliance with OIML R111
  • Supplied individually or as a set, in a plastic case or a wooden box
  • Specifications

    Specifications

    1 to 50 g
    For use with Weighing Instruments of Medium Accuracy Class III and Ordinary Accuracy Class IIII
    Product Suggestions

    Product Suggestions

    Videos
    SDS
    Documents

    Documents

    Certificates
    Special Offers

    Special Offers