Learn More
Abeba™ Clean Room 3620 Shoes
Special ESD clean room shoes.
Brand:  Abeba™ 3620/46
 View more versions of this product
Product Code. 15610388
  • 105.00 EUR / 1 pair
Estimated Shipment: 07-02-2025
Description
  • CE EN ISO 20347:2012 OB A, E SRA
  • Microfiber
  • White
  • ESD dissipative according to DIN EN 61340
  • Fixed gaitor, cleanroom textile
Specifications
Specifications
CE EN ISO 20347:2012 OB A, E SRA
Microfiber
3620
White
46