Learn More
Cole-Parmer™ AccTech™ OpticPad™ Cleaning Pads
Product Code. 11720579 Shop All Cole Parmer Products
Click to view available options
For Use With (Application):
For Nonabrasive surface cleaning and polishing
Use these cleaning pads for general optical cleaning applications. AccTech™ OpticPad™ Cleaning Pads are ideal for nonabrasive surface cleaning and polishing.
100 pads per box
Specifications
Product Type Cleaning Pad
For Use With (Application) For Nonabrasive surface cleaning and polishing
Quantity 100/Pk.
compliance-icons compliance-icons
Product Identifier
  • opticpad
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : propan-2-ol, ammonium hydroxide