Learn More
Labconco™ Clear Drying Chamber for FreeZone™ Freeze Dryers
Shop All Labconco Corporation Products
Click to view available options
Size:
Short
Tall
Height (English):
15.3 in.
16.8 in.
23 in.
24.6 in.
Unit Size:
Each
Description
Clear chamber provides visibility for bulk freeze drying.
- 12.0 in. dia. acrylic chamber fits all FreeZone™ Freeze Dryers
- Provides visibility for bulk freeze drying
- Includes clear acrylic lid with gasket, vacuum release valve and pass-through for electrical cords
- Will fit 10.12 in. or 14.0 in. diameter condensers
- 8 in. diameter opening to collector on bottom of chamber
Specifications
Specifications
| Type | Drying Chamber |
| No. of Shelves | 5 |
| Color | Clear |
| Material | Acrylic |
| Size | Tall |
| For Use With (Equipment) | FreeZone™ Freeze Dryers |
| Includes | 8 Valves |
| Diameter (English) | 17.4 in. |
| Height (English) | 23 in. |
| Shipping Weight (English) | 25 lb. |
| Show More |