Learn More
Invitrogen™ CNKSR3 Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA5113045
Product Code. 17160904
  • 461.00 EUR / 100µg
Description
CNKSR3 Polyclonal Antibody for Western Blot, ICC/IF, ELISA
CNKSR3 may be involved in transepithelial sodium transport. Regulates aldosterone-induced and ENaC-mediated sodium transport possibly through regulation of the ERK pathway.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CNKSR3
Polyclonal
Unconjugated
CNKSR3
6820402C05; BC024086; CNK homolog protein 3; CNK3; CNKSR family member 3; Cnksr3; Connector enhancer of kinase suppressor of ras 3; connector enhancer of KSR 3; KOX14; MAGI1; maguin-like protein; membrane associated guanylate kinase interacting protein-like 1; membrane associated guanylate kinase, WW and PDZ domain containing 1; membrane-associated guanylate kinase-interacting protein-like 1; parturition-related protein 4; Prp4; ZNF21
Rabbit
Protein G
RUO
154043, 215748
-20°C or -80°C if preferred
Liquid
ELISA, Western Blot, Immunocytochemistry
1 mg/mL
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Q6P9H4, Q8BMA3
CNKSR3
Recombinant Human Connector enhancer of kinase suppressor of ras 3 protein (301-555AA).
100 μg
Primary
Human, Mouse
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions