missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(II) Chloride Solution, 5.95%, Red, Chem-Lab

Product Code. 11927433
Click to view available options
Quantity:
100 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

Used as an indicator for water in desiccants. Cobalt(II) Chloride Solution, 5.95%, Red, Chem-Lab is an inorganic compound of cobalt and chlorine, supplied as a hexahydrate. Reagent according to Eur. Ph. 4 (2.2.2).

Cobalt(II) chloride hexahydrate is one of the most commonly used cobalt compounds in the lab. It is a red color at room temperature, but becomes blue at higher temperatures. Because of the ease of the hydration/dehydration reaction, and the resulting color change, cobalt chloride is used as an indicator for water in desiccants. Niche uses of cobalt chloride include its role in organic synthesis and electroplating objects with cobalt metal.

Specifications

Melting Point 0°C
CAS 7791-13-1
Color Red
pH <1
Physical Form Liquid
Quantity 100 mL
Molecular Formula Cl2CoH12O6
MDL Number MFCD00149652
Synonym Cobalt dichloride
Solubility Information Soluble in Water
InChI Key GFHNAMRJFCEERV-UHFFFAOYSA-L
SMILES O.O.O.O.O.O.[Cl-].[Cl-].[Co++]
IUPAC Name λ²-cobalt(2+) hexahydrate dichloride
Molecular Weight (g/mol) 237.92
Formula Weight 237.90g/mol
Concentration or Composition (by Analyte or Components) 5.95%
Odor Odorless
Density 1.01g/mL
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Cobalt(II) chloride 5.95% solution (Red sol.)
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • Carcinogenicity Category 1B
  • Germ cell mutagenicity Category 2
  • Reproductive toxicity Category 1B
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H341-Suspected of causing genetic defects.
  • H350-May cause cancer.
  • H360-May damage fertility or the unborn child.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : cobalt chloride, hydrochloric acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'