Learn More
Bollé Safety Cobra ESP Safety Glasses
Offers 180°; vision and perfect optical quality. Bollé™ COBRA Wrap-around Safety Glasses are the essential all rounder with panoramic visual field.
Brand: Bollé Safety COBESP
This item is not returnable.
View return policy
Description
- One product = two uses
- Upper protection
- Panoramic vision
- Non-slip bridge
- Comfortable, non-slip straight temples
- Temple or strap version with interchangeable foam
- Pivoting temples
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Frame: EN 166 BT CE; Lens marking: 5-1.4 1 BT CE; Chemical risk - Liquid droplets - 3 (Goggle, adapted spectacles with strap or face shield mandatory); Mechanical risk - Low energy impact - F; Mechanical risk- High speed particles at extreme temperatures | |
COBRA wrap around safety glasses | |
B-ESP filters; Scratch-resistant, anti-fog (ASAF) | |
25 g | |
Wraparound, non-slip bridge |
Anti-fog, Scratch-resistant | |
Black | |
1 Ea. | |
Panoramic vision |