missing translation for 'onlineSavingsMsg'
Learn More
Learn More
COD, Chloride Standard Solution, WTW™
Click to view available options
Quantity:
100 mL
Specifications
Specifications
Physical Form | Liquid |
Concentration | COD 250mg/L, Chloride 125mg/L |
Solution Contains | COD, Chloride |
Color | Colorless |
Quantity | 100 mL |
Solubility | Soluble in water |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'