Learn More
SCHOTT AG Lighting and Imaging™ Color Filter for Spotlight
Color Filter for use with EasyLED single or double Spotlight Plus. Available in Red, Blue, Green, Yellow or Halogen and a polarization filter.
Brand:  SCHOTT AG Lighting and Imaging™ 600660
Additional Details : Weight : 5.00000kg
 View more versions of this product
Product Code. 15723052
  • 47.50 EUR / 1 unit
Estimated Shipment: 20-11-2024
to see stock.
Add to basket
Description
Description
  • Color filters for EasyLED single or double Spotlight Plus
  • Use for fixing at spot light with thread.
  • Available colors: red, blue, green, yellow and halogen.
  • Polarization filter available.
Specifications
Specifications
Color Filter
Convert neutral white LED light to blue light
EasyLED Spotlight Plus (Single or Double)
Blue
Stereo Microscopy Illumination