Learn More
Fisherbrand™ Combs for MultiSUB Maxi Horizontal Gel Electrophoresis Unit, 0.75mm Thick
For use in preparatory electrophoresis and high-throughput screening.
31.75 EUR - 34.30 EUR
Specifications
For Use With (Equipment) | MultiSUB Maxi-Gel Electrophoresis Unit |
---|---|
Type | Comb |
Quantity | 1 Each |
Thickness (Metric) | 0.75 mm |
Product Code | Brand | No. of Wells | Volume (Metric) Sample | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | No. of Wells | Volume (Metric) Sample | Price | Quantity & Availability | ||||
11813453
|
Fisherbrand™
MS20-36-0.75 |
36 | 11 μL |
31.75 EUR
Each |
Estimated Shipment: 20-05-2025
Log in to see stock. |
||||
11853453
|
Fisherbrand™
MS20-4-0.75 |
4 Prep, 2 Marker | 115 μL |
31.75 EUR
Each |
Estimated Shipment: 20-05-2025
Log in to see stock. |
||||
11873443
|
Fisherbrand™
MS20-30-0.75 |
30 | 13 μL |
32.40 EUR
Each |
Estimated Shipment: 20-05-2025
Log in to see stock. |
||||
11853433
|
Fisherbrand™
MS20-2-0.75 |
2 Prep, 2 Marker | 236 μL |
31.75 EUR
Each |
Estimated Shipment: 20-05-2025
Log in to see stock. |
||||
11813433
|
Fisherbrand™
MS20-16-0.75 |
16 | 30 μL |
31.75 EUR
Each |
Estimated Shipment: 20-05-2025
Log in to see stock. |
||||
11833423
|
Fisherbrand™
MS20-1-0.75 |
1 Prep, 1 Marker | 506 μL |
31.75 EUR
Each |
Estimated Shipment: 20-05-2025
Log in to see stock. |
||||
11843423
|
Fisherbrand™
MS20-10-0.75 |
10 | 54 μL |
33.45 EUR
Each |
Estimated Shipment: 20-05-2025
Log in to see stock. |
||||
11863453
|
Fisherbrand™
MS20-40MCSS-0.75 |
40 | 8 μL |
31.75 EUR
Each |
Estimated Shipment: 20-05-2025
Log in to see stock. |
||||
11833463
|
Fisherbrand™
MS20-50-0.75 |
50 | 8 μL |
31.75 EUR
Each |
Estimated Shipment: 20-05-2025
Log in to see stock. |
||||
11863433
|
Fisherbrand™
MS20-20MC-0.75 |
20 | 20 μL |
31.75 EUR
Each |
Estimated Shipment: 22-04-2025
Log in to see stock. |
||||
11833443
|
Fisherbrand™
MS20-25-0.75 |
25 | 16 μL |
34.30 EUR
Each |
Estimated Shipment: 20-05-2025
Log in to see stock. |
||||
Description
- Recommended for high resolution gels and publication quality data
- For tightly resolved bands
- Height adjustable to give user full control over well depth and sample-loading volume
- Rigid back which prevents heat-induced warping
- Reversible loading guides sit directly above each well to provide a convenient loading template for single and multichannel pipettes
- 20-sample and 40-sample combs are Multichannel pipettor compatible
Specifications
MultiSUB Maxi-Gel Electrophoresis Unit | |
Comb | |
1 Each | |
0.75 mm |