missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Complement C3a Antibody (K13/16-5.7) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-05122
This item is not returnable.
View return policy
Description
Complement C3a Monoclonal specifically detects Complement C3a in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Sandwich ELISA Capture.
Specifications
| Complement C3a | |
| Monoclonal | |
| 1.0 mg/mL | |
| Western Blot 1:100-1:2000, ELISA 1:10000, Immunohistochemistry 1:150, Immunocytochemistry/Immunofluorescence 1:75, Immunohistochemistry-Paraffin 1:150, Sandwich ELISA Capture | |
| C3 | |
| Human C3a | |
| 100 μg | |
| Primary | |
| Specific for human C3, C3a and C3a (desArg) and does not cross-react with C4a or C5a. | |
| Store at 4C. Do not freeze. | |
| IgG1 κ |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| K13/16-5.7 | |
| Unconjugated | |
| AHUS5, ARMD9, ASP, C3 and PZP-like alpha-2-macroglobulin domain-containing protein 1, complement C3, complement component 3, complement component C3, CPAMD1acylation-stimulating protein cleavage product | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 718 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction