Learn More
Agilent BioTek Complete Direct Drain Waste Systems
Drains into sink instead of bottles
Specifications
Includes | Direct drain module (115V/230V), standard or high-flow vacuum pump, tubing, holder for two 4L bottles |
---|---|
For Use With (Equipment) | Agilent BioTek 405 Series Microplate Washer System, Agilent BioTek 406 Series Microplate Washer System |
Description
- Compatible with Agilent BioTek 405 and 406 Series Microplate Washer Systems
- Eliminates the need of attending to and emptying waste bottles
- Saves space on the workbench
Specifications
Direct drain module (115V/230V), standard or high-flow vacuum pump, tubing, holder for two 4L bottles | |
Agilent BioTek 405 Series Microplate Washer System, Agilent BioTek 406 Series Microplate Washer System |