missing translation for 'onlineSavingsMsg'
Learn More

Compound E, MedChemExpress

Product Code. p-200098964
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
Unit Size:
10mg
1mg
25mg
5mg
This item is not returnable. View return policy

Product Code. 16473406

Brand: Medchem Express HY14176

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Compound E is a γ-secretase inhibitor. Compound E bloks β-amyloid(40), β-amyloid(42), and Notch γ-secretase cleavage with IC50s of 0.24, 0.37, 0.32 nM, respectively.

Compound E is a γ-secretase inhibitor. Compound E bloks β-amyloid(40), β-amyloid(42), and Notch γ-secretase cleavage with IC50s of 0.24, 0.37, 0.32 nM, respectively.

Specifications

Chemical Name or Material Compound E
CAS 209986-17-4
Color White
Purity Grade Notes Research
Molecular Formula C27H24F2N4O3
Quantity 25 mg
Synonym γ-Secretase-IN-1
Solubility Information DMSO : 115 mg/mL (234.45 mM; ultrasonic and warming and heat to 60°C)
SMILES FC1=CC(CC(N[C@@H](C)C(N[C@@H]2C(N(C)C(C=CC=C3)=C3C(C4=CC=CC=C4)=N2)=O)=O)=O)=CC(F)=C1
Molecular Weight (g/mol) 490.5
Formula Weight 490.5
Percent Purity 98.75%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.