missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SI Analytics™ Conductivity Cell with 3m Fixed Cable
SI Analytics™ Conductivity Cell with 3m Fixed Cable is constructed of a graphite sensor and plastic shaft.
Brand: SI Analytics™ 285106148
This item is not returnable.
View return policy
Description
- 4 pole cell
- Plastic shaft
- 3m cable with 8 pole plug
Specifications
4 pole cell | |
15.3 mm | |
120 mm | |
3 m |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction