missing translation for 'onlineSavingsMsg'
Learn More

Conductivity Resistivity Standard Solution, 1330μS/cm, Reagecon™

Product Code. 10388547 Shop All Reagecon Products
Click to view available options
Quantity:
500 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

Quantity 500 mL
Color Colorless
Odor Characteristic
Density 1.00091g/cm3 at 20°C
Conductivity 1330μS/cm
Solubility Completely miscible
Physical Form Liquid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.