missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Congo Red, indicator grade

Product Code. 11425737
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 11425737

Brand: Thermo Scientific Chemicals B24310.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 573-58-0 | C32H22N6Na2O6S2 | 696.664 g/mol

Congo red is useful for staining and visual detection of amyloid deposits. It inhibits the accumulation of the scrapie-associated, protease-resistant isoform of protein PrP without affecting the metabolism of the normal isoform.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Congo red is useful for staining and visual detection of amyloid deposits. It inhibits the accumulation of the scrapie-associated, protease-resistant isoform of protein PrP without affecting the metabolism of the normal isoform.

Solubility
Soluble in water 25 g/L.

Notes
Store away from oxidizing agents, acids, reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 573-58-0
Molecular Formula C32H22N6Na2O6S2
Molecular Weight (g/mol) 696.664
MDL Number MFCD00004028
InChI Key IQFVPQOLBLOTPF-UHFFFAOYSA-L
Synonym C.I. 22120
PubChem CID 11313
ChEBI CHEBI:34653
IUPAC Name disodium;4-amino-3-[[4-[4-[(1-amino-4-sulfonatonaphthalen-2-yl)diazenyl]phenyl]phenyl]diazenyl]naphthalene-1-sulfonate
SMILES C1=CC=C2C(=C1)C(=CC(=C2N)N=NC3=CC=C(C=C3)C4=CC=C(C=C4)N=NC5=C(C6=CC=CC=C6C(=C5)S(=O)(=O)[O-])N)S(=O)(=O)[O-].[Na+].[Na+]

Specifications

Melting Point >360°C
Beilstein 3894858
Merck Index 14,2498
Solubility Information Soluble in water 25g/L.
Formula Weight 696.67
Grade Indicator
Odor Odorless
Quantity 100 g
Chemical Name or Material Congo Red
compliance-icons
Product Identifier
  • Congo Red
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 1B
  • Reproductive toxicity Category 2
Hazard Statement
  • H350-May cause cancer.
  • H361d-Suspected of damaging the unborn child.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : C.I. Direct Red 28

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.