Learn More
SCAT Connectors for HPLC
Safe connection of tubes of different sizes: directly and without technical aids. For safe collection of liquid HPLC waste, and many other applications in the laboratory!
Brand: SCAT 107822
10.30 EUR valid until 2024-12-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
SCAT™ connectors are available in the following versions:
- Conical connectors
- Y-connectors
- Tube connectors
- Capillary connectors
Specifications
Conical Connector | |
Polypropylene | |
HPLC/UHPLC Systems |
4 to 8 mm, 8 to 12 mm | |
Tubing |