Learn More
WTW™ ConOx™ -1.5 Combined Oxygen/Conductivity Sensor
Brand: WTW™ 401010
This item is not returnable.
View return policy
Description
Specifications
Combined Oxygen/Conductivity Sensor | |
Epoxy, Graphite | |
ConOx-1.5 |
1.5 m | |
1.5m L Cable, with 3 exchange membrane heads, cleaning solution, conductivity standard (1413μS/cm), abrasive foil, OxiCal™ CX calibration vessel, electrolyte |