Learn More
Corning™ Extension Collar for CoolBox™ 2XT System
BIOPHARMA
Product Code. p-6960009
Click to view available options
Color:
Green
Orange
Pink
Purple
Unit Size:
Each
Lid closure not possible with addition of extension collar
Product Code. 15522861
Brand: Corning™ 432087
Only null left
Add to basket
Add to basket
Lid closure not possible with addition of extension collar
For use with CoolBox™ 2XT System. Corning™ Extension Collar is required when using taller CoolRack™ tube modules and tall tubes in CoolBox system.
For use with CoolBox XT and CoolBox 2XT to extend the height when using tall tube modules. The collar is magnetized and easily adheres to the unit base.
TRUSTED_SUSTAINABILITY
Specifications
Description Extension Collar
Type Magnetized Extension Collar
Color Purple
No. per Pack 1/Pk.
No. per Case 1/Cs.
For Use With (Application) To extend height of CoolBox™ ice-free cooling systems
For Use With (Equipment) CoolBox 2XT System
Dimensions (L x W x H) 21.0 x 14.3 x 7.0 cm
Dimensions (L x W x H) Exterior 26.5 x 20.0 x 7.0 cm