Learn More
Copper based low temperature water gas shift catalyst, HiFUEL™ W220, Thermo Scientific™
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
Unit Size:
1kg
2.5kg
500g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Spécification
Spécification
| Chemical Name or Material | Copper based low temperature water gas shift catalyst, HiFUEL W220 |
| Color | Black |
| Appearance | Black pellets |
| Quantity | 500 g |
| Form | Pellet |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu