Learn More
Copper gauze, 140 mesh woven from 0.056mm (0.0022in) dia wire, Thermo Scientific Chemicals
Product Code. 15467265
Click to view available options
Quantity:
75 x 75 mm
150 x 150 mm
300 x 300 mm
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 7440-50-8
Molecular Formula Cu
Molecular Weight (g/mol) 63.55
MDL Number MFCD00010965
InChI Key RYGMFSIKBFXOCR-UHFFFAOYSA-N
Synonym cuprum, cobre, cuivre, blister, cathode, bronze, powder, anode, precipitates, kupfer
PubChem CID 23978
ChEBI CHEBI:30052
IUPAC Name copper
SMILES [Cu]
Specifications
Quantity 300 x 300 mm
Mesh Size 140 Mesh
Odor Odorless
Form Wire Cloth
Chemical Name or Material Copper gauze, 140 mesh woven from 0.056mm (0.0022 in.) dia. wire