missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper(I) Cyanide, purum p.a, ≥99.0% (KT), Honeywell™
Click to view available options
Quantity:
50 g
250 g
1 kg
Packaging:
Poly bottle
Description
- Products with a guarantee certificate and/or suitable for the stated analytical application.
Specifications
Specifications
Melting Point | 473°C |
Color | White |
Physical Form | Powder |
Quantity | 1 kg |
Linear Formula | CuCN |
UN Number | UN1587 |
Beilstein | 3587244 |
Formula Weight | 89.56g/mol |
Percent Purity | ≥99.0% (KT) |
Odor | Odorless |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction