missing translation for 'onlineSavingsMsg'
Learn More

Copper(I) Cyanide, purum p.a, ≥99.0% (KT), Honeywell™

Product Code. 15676020
Click to view available options
Quantity:
50 g
250 g
1 kg
Packaging:
Poly bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Purum p.a.,≥99.0% (KT)

  • Products with a guarantee certificate and/or suitable for the stated analytical application.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 544-92-3
Molecular Formula CCuN
Molecular Weight (g/mol) 89.564
MDL Number MFCD00010975
InChI Key DOBRDRYODQBAMW-UHFFFAOYSA-N
Synonym copper i cyanide, cupricin, copper +1 cyanide, copper cyanide cu cn, cyanocopper, rcra waste number p029, cucn, rcra waste no. p029, cyano copper
PubChem CID 11009
IUPAC Name copper(1+);cyanide
SMILES [C-]#N.[Cu+]

Specifications

Melting Point 473°C
Color White
Physical Form Powder
Quantity 250 g
Linear Formula CuCN
UN Number UN1587
Beilstein 3587244
Formula Weight 89.56g/mol
Percent Purity ≥99.0% (KT)
Odor Odorless
Packaging Poly bottle
Density 2.92g/mL at 25°C
Chemical Name or Material Copper(I) cyanide
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.