missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper Solution for ICP, 1g/L, HNO3 Matrix, Fisher Chemical™
Click to view available options
Quantity:
100 mL
500 mL
1000 mL
Specifications
Specifications
Concentration | 1μg/mL±0.002g/L |
Packaging | Bottle |
Quantity | 1000 mL |
Grade | EU Ph Ref: 1022300 |
Solvent or Matrix | Methanol |
Chemical Name or Material | Copper- Cu |
Safety and Handling
Product Identifier
- Single-element Standard Solution for ICP. Copper (Cu) 1000mg/l in HNO3 2%
- LONG-TERM AQUATIC HAZARD Chronic 2
- H411-Toxic to aquatic life with long lasting effects.
- P273-Avoid release to the environment.
- P391-Collect spillage.
- P501-Dispose of contents/container to .
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'