Learn More
JULABO CORIO CP Refrigerated/Heating Circulator
Click to view available options
Refrigerant:
R290
R449A
Model:
CP-310F
CP-450F
Unit Size:
Each
Description
- Models for internal and external applications
- Bright, white, easy to read display
- Very quiet
- Easy pump change-over between internal and external circulation
- External pump connections
- Powerful and infinitely adjustable pressure pump
- Bath lid and drain tap included
- Removable ventilation grid
- Refrigeration unit without side vents
- Unique early warning system for low liquid level
- Class III (FL) according to DIN 12876-1
Specifications
Specifications
| Capacity (Metric) | 3 to 4 L |
| Certifications/Compliance | CE |
| Communications | USB, RS232 |
| Cooling Capacity | 300 W |
| Flow Rate | 8 to 27 L/min. |
| Pump Type | Centrifugal |
| Temperature Range | -30°C to +200°C |
| Type | Circulator |
| Model | CP-310F |
| Refrigerant | R449A |
| Show More |