Learn More
Corning™ DeckWorks™ Bulk Pipet Tips
Product Code. 11373635 Shop All Corning Products
Click to view available options
Volume (Metric):
0.1 to 10 μL
1 to 200 μL
100 to 2000 μL
Color:
Natural
Yellow
Unit Size:
Pack of 1000
Pack of 10000
Pack of 768
Pack of 7680
Product Code. 11373635
missing translation for 'mfr': Corning™ 4111
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Compatible with DeckWorks racks and packaged in resealable polyethylene bags
Corning DeckWorks bulk pipet tips are compatible with DeckWorks racks and packaged in resalable polyethylene bags. Tip sizes include 10 μL, 200 μL, and 1000 μL and offer ­maximum savings.
TRUSTED_SUSTAINABILITY
Specifications
Volume (Metric) 1 to 200 μL
Sterility Non-sterile
Color Natural
For Use With (Application) Help reduce the production and waste of plastic materials
Tip Style Round
Packaging Bulk
Disposable Single-use
For Use With (Equipment) DeckWorks Racks
Graduated Yes
Product Line DeckWorks