Learn More
Corning™ Human Umbilical Vein Endothelial Cells (HUVEC-2)
Shop All Corning Products
missing translation for 'orderingAttributeHoverText'
Quantity:
>5 x 105 Cells
missing translation for 'unitSize'
Each
Description
Corning™ HUVEC-2 cells are qualified for use in Corning™ BioCoat™ Angiogenesis Systems: Endothelial Cell Migration. The cells may be used in Corning BioCoat Endothelial Cell Invasion and Tube Formation assays. Packaged in individual cryogenic vials containing >5x105 cells.
- Excellent viability
- Each vial of Corning™ HUVEC-2 cells contain 5x105 cells that have been cryopreserved at passage number 2 in medium containing 10% DMSO.
- Lot-to-lot consistency
- Tested for the ability to produce a robust migratory response in the presence of VEGF and FBS.
- Prescreened for Angiogenesis studies
- Qualified for use in Corning™ BioCoat Angiogenesis Systems: Endothelial Cell Migration. May also be used in Corning™ BioCoat Endothelial Cell Invasion and Tube Formation assays.
- Positive immunohistochemical staining for von Willebrand factor (vWf) and CD31 antigen
- Positive Dil-Ac-LDL uptake
- Negative mycoplasma, HIV-1, hepatitis B and C, bacteria, yeast, and fungi
Specifications
Specifications
| For Use With | Corning™ BioCoat™ Angiogenesis System |
| For Use With (Application) | Used as model for studying endothelial differentiation and modulation of endothelial tube formation by antiangiogenic agents |
| Product Type | Human Umbilical Vein Endothelial Cells |
| Quantity | >5 x 105 Cells |
| Source | Human Umbilical Vein |