Learn More
Corning™ Lambda Plus Single-Channel Pipettor, Adjustable, Autoclavable
GREENER_CHOICE
Product Code. 11317431 Shop All Corning Products
Click to view available options
Volume (Metric):
0.1 to 2 μL
0.5 to 10 μL
10 to 100 μL
100 to 1000 μL
2 to 20 μL
20 to 200 μL
Accuracy:
±1 to ±0.6%
±1.2 to ±0.6%
±1.6 to ±0.8%
±12 to ±1.5%
±4 to ±0.5%
±4 to ±0.8%
Compatible Tips:
10μL Tip
1000μL Pipette tip
200μL Tip
missing translation for 'unitSize'
Each
Product Code. 11317431
missing translation for 'mfr': Corning™ 4072
Only null left
Add to basket
Add to basket
Designed for universal fit with all common brands of pipet tips
Engineered to provide the highest levels of accuracy, reproducibility, and comfort
Available in single-, eight-and twelve-channel configurations, all Lambda Plus pipettors feature smooth plunger movement and extremely low pipetting forces. Volume can be set by turning either the thumb wheel or the plunger button, and a locking mechanism prevents accidental change. The Lambda Plus pipettors have been designed to maintain their calibration, even after repeated use and autoclaving. A key is included with each pipettor for easy in-house calibration.
Features
  • Factory calibrated to strict ISO 8655 standards-3-year warranty
  • Fully autoclavable and UV resistant
  • Designed for universal fit with all common brands of pipet tips
  • Adjustable tip ejector
Services
3 year
TRUSTED_SUSTAINABILITY
Specifications
Accuracy ±4 to ±0.8%
Certifications/Compliance ISO8655
Compatible Tips 200μL Tip
Autoclavable Autoclavable
For Use With 200 μL Tip
Number of Channels 1
Product Line Lambda Plus
Type Single-Channel Pipettor
Volume (Metric) 2 to 20 μL
For Use With (Equipment) 200μL Tip
Includes Key is included with each pipette for easy in-house calibration
UV Monitoring Yes
Warranty 3 Years