Learn More
Corning™ Plain Reusable Short Stem Plastic Funnels
Product Code. p-4901455 Shop All Corning Products
Click to view available options
Diameter (Metric) Stem:
1.5 mm
4 mm
6 mm
7 mm
11 mm
15 mm
Height (Metric):
45 mm
65 mm
85 mm
110 mm
180 mm
220 mm
Length (Metric) Stem:
25 mm
35 mm
43 mm
55 mm
90 mm
95 mm
Unit Size:
Pack of 12
Pack of 24
Product Code. 17263554
Brand: Corning™ 6120P30
Only null left
Add to basket
Add to basket
Durable polypropylene funnels are translucent and allow safe and easy transfers of solids or liquids from one vessel to another.
Allows safe and easy transfer of solids or liquids from one vessel to another.
TRUSTED_SUSTAINABILITY
Specifications
Diameter (Metric) Stem 1.5 mm
Material Polypropylene
Height (Metric) 45 mm
Description Constructed of Durable translucent polypropylene
Length (Metric) Stem 25 mm