Learn More
Corning™ Reusable Plastic Erlenmeyer Flasks with Red Graduations
Translucent polymethylpentene; with polypropylene screw cap
Brand:  Corning™ 4990P-500
 View more versions of this product
Product Code. 17233554
  • 116.48 EUR / Pack of 6
Estimated Shipment: 26-05-2025
to see stock.
Explore more special offers
Description
Translucent polymethylpentene flasks are manufactured with red graduations for reference. The flasks have a screw neck and are supplied with a PP cap.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Polypropylene
Polypropylene
Narrow Mouth
1/Pk.
500 mL
100 mL
6/Cs.
Screw Cap