Learn More
Corning™ Universal Fit Pipet Tips: Racked, Nonsterile
Shop All Corning Products
Click to view available options
Volume (Metric):
1 to 200 μL
100 to 1000 μL
Color:
Blue
Natural
Yellow
Unit Size:
Pack of 1000
Pack of 960
Description
Designed to assure a reliable fit with major brand pipetters
- Corning universal fit tips are designed to provide a reliable fit with most major brand pipettors. (A Pipet Tip Compatibility Guide can be requested or downloaded from the Corning website).
- Beveled orifice helps ensure accurate fluid delivery.
- 1-200 μL universal fit tips are graduated at the 250 μL, 500 μL, and 1,000 μL volumes.
- Select from three packaging options: Stack rack tips feature a stack of five racks, each containing 96 tips, for a total of 480 tips in a space-saving design. Bulk packed tips are nonsterile and economical.
- RNase-/DNase-free.
- Nonpyrogenic.
Specifications
Specifications
| Volume (Metric) | 100 to 1000 μL |
| Sterility | Non-sterile |
| Color | Blue |
| Format | Rack |
| Tip Style | Round |
| Packaging | Rack |
| For Use With (Equipment) | Universal Fit |
| Graduated | Yes |
| Material | Polypropylene |