Learn More
Emma Safety Footwear Corona Safety Shoes
Washable microfiber uppers are treated to repel all kinds of dirt, germs and bacteria. Highly breathable, comfortable shoe for all day wear. Energy returning soles prevent fatigue and keep the wearer fresh, alert, foot sure and steady even in the event of a spillage.
Brand: Emma Safety Footwear MM54454039
This item is not returnable.
View return policy
Description
- S2-Safety shoe for wet working conditions
- GripForce™ Easy Twist Outsole
- Microfiber construction
- Chrome-free lining
- Hydro-Tec™ Sanitized Silver
Specifications
EN ISO 20345:2011 | |
White | |
Hydro-Tec™ Sanitized Silver | |
39 |
Laces | |
Corona | |
Microfiber | |
Steel |