Learn More
Corning™ Costar™ 8-Pette™ and 12-Pette™ Multichannel Pipetters
Shop All Corning Products
Click to view available options
Number of Channels:
8
12
Unit Size:
Each
Description
These20-200 μL multi-channel pipettors feature a unique, ergonomic trigger-style aspiration and dispense control mechanism designed to reduce thumb fatigue during repetitive pipetting.
• Volume adjusts with a vernier-scale spindle
• Pipettors are entirely autoclavable
Order Info
Compatible with all Fisherbrand™ universal-fit tips.
Specifications
Specifications
| Autoclavable | Autoclavable |
| Number of Channels | 12 |
| Product Line | Costar 8-Pette and 12-Pette |
| Sterility | Non-sterile |
| Type | Pipette |
| Volume (Metric) | 20 to 200 μL |