Learn More
Corning™ Costar™ Snap Cap Microcentrifuge Tubes
Shop All Corning Products
Click to view available options
Color:
Assorted
Natural
Capacity (Metric):
0.65 mL
1.7 mL
2 mL
Packaging:
1000/Pk.
500/Pk.
5000/Pk.
Unit Size:
Pack of 1000
Pack of 500
Pack of 5000
Description
- Supplied nonsterile and are autoclavable.
- External graduations and frosted writing spot for easy sample identification.
- Positive seal design allows repeated opening and closing.
- Flat cap surface for convenient labeling.
- Withstands a maximum RCF of 17,000 x g.
- RNase-/DNase-free.
Specifications
Specifications
| Cap Style | Snap Cap |
| Certifications/Compliance | RNase, DNase-free |
| Closure Color | Natural |
| Closure Material | Polypropylene |
| Autoclavable | Autoclavable |
| Capacity (Metric) | 0.65 mL |
| Color | Natural |
| Material | Polypropylene |
| No. per Pack | 500/Pk. |
| Packaging | 1000/Pk. |
| Show More |