Learn More
Sperian™ Honeywell™ Cotton/Elasthane White Gloves
Click to view available options
Size:
6/8
8/10
Unit Size:
12 pairs
Pack of 12
Description
- Industrial cut, edged border
- Ambidextrous and thus more cost-effective
- Cotton interlock ensures the glove's mechanical performance while absorbing hand perspiration for added comfort
- Tight knit gives the glove perfect support and limits the penetration of dirt
- Ribbed at the wrist to ensure a good level of wrist comfort and eliminates the risk of it getting caught
Specifications
Specifications
| Classification | Class 5 |
| Construction | Jersey Knit |
| Hand | Ambidextrous |
| Material | Cotton, Elasthane |
| Product Type | Gloves |
| Size | 6/8 |
| For Use With (Application) | Finishing operations in mechanical, clock-making, jewelery, photography. Handlings requiring a clean glove without any plush. |
| Quantity | 12/Pk. |
| Length (Metric) | 19.5-21 cm |