Learn More
Fisherbrand™ Cotton Linter Antibiotics Testing Paper
Tests antibiotic sensitivity of bacteria.
Brand:  Fisherbrand™ 0905A00001
Product Code. 11780096
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • Thick absorbent paper
  • 100% cotton linter fibre
  • Weight:330g/m2
  • Thickness: 0.9mm
  • Water absorption: 215mL
Applications:
  • Impregnation of antibiotics for testing resistance of pathogenic bacteria
Specifications
Specifications
Impregnation of antibiotics for testing the resistance of pathogenic organisms
Cotton Linter
1000 Pack
6 mm