missing translation for 'onlineSavingsMsg'
Learn More

Coumalic acid, 97%

Product Code. p-7028290
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11497196

Brand: Thermo Scientific Alfa Aesar B25521.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 500-05-0 | C6H4O4 | 140.09 g/mol

Coumalic acid undergoes thermal reaction with 1,3-butadiene to yield dimethyl tricycle[3.2.1.02,7]oct-3-ene-2,4-dicarboxylate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Coumalic acid undergoes thermal reaction with 1,3-butadiene to yield dimethyl tricycle[3.2.1.02,7]oct-3-ene-2,4-dicarboxylate.

Solubility
Very soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 500-05-0
Molecular Formula C6H4O4
Molecular Weight (g/mol) 140.09
MDL Number MFCD00006644
InChI Key ORGPJDKNYMVLFL-UHFFFAOYSA-N
Synonym coumalic acid, 2-oxo-2h-pyran-5-carboxylic acid, cumalic acid, 2h-pyran-5-carboxylic acid, 2-oxo, 2-oxopyran-5-carboxylic acid, unii-ob1jpy343g, 2-pyrone-5-carboxylic acid, alpha-pyrone-5-carboxylic acid, .alpha.-pyrone-5-carboxylic acid, ob1jpy343g
PubChem CID 68141
SMILES OC(=O)C1=COC(=O)C=C1

Specifications

Melting Point ∼208°C (decomposition)
Boiling Point 218°C (120 mmHg)
Quantity 5 g
Beilstein 119747
Merck Index 14,2557
Solubility Information Very soluble in water.
IUPAC Name 6-oxopyran-3-carboxylic acid
Formula Weight 140.1
Percent Purity 97%
Chemical Name or Material Coumalic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.