missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CPOX Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12211-1-AP-20UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene is the sixth enzyme of the heme biosynthetic pathway. The encoded enzyme is soluble and found in the intermembrane space of mitochondria. This enzyme catalyzes the stepwise oxidative decarboxylation of coproporphyrinogen III to protoporphyrinogen IX, a precursor of heme. Defects in this gene are a cause of hereditary coproporphyria (HCP).Specifications
CPOX | |
Polyclonal | |
Unconjugated | |
CPOX | |
Coprogen oxidase, Coproporphyrinogen oxidase, Coproporphyrinogenase, COX, CPO, CPOX, CPX, HCP | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
12892, 1371, 304024 | |
-20°C | |
Liquid |
Immunoprecipitation, Immunohistochemistry (Paraffin), Western Blot | |
0.29 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P36551, P36552, Q3B7D0 | |
CPOX | |
CPOX Fusion Protein Ag2858 | |
20 μL | |
Primary | |
Rat, Mouse, Human | |
Antibody | |
IgG |