missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CRALBP Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA583588
This item is not returnable.
View return policy
Description
Immunogen sequence: TTKDHGPVFG PCSQLPRHTL QKAKDELNER EETREEAVRE LQEMVQAQAA SGEELAVAVA ERVQEKDSGF FLRFIRARKF NVGRAYELLR GYVNFRL Highest antigen sequence indentity to the following orthologs: Rat - 92%, Mouse - 91%.
Cellular retinaldehyde-binding protein (CRALBP) plays an important role in the regeneration of 11-cis-retinal for use in rod visual pigments such as opsin and rhodopsin. Once 11-cis-retinal is photoisomerized in the rod outer segment, it is converted to all-trans-retinal and further modified into all-trans-retinol. All-trans-retinol then diffuses into the retinal pigment epithelium (RPE) to be converted back to 11-cis-retinol and further oxidized into 11-cis-retinal (both by CRALBP). Genetic mutations involving CRALBP's lack of function have been linked to visual disease such as bothnia dystrophy, retinitis punctata albescens, retina pigmentosa, and Newfoundland rod-cone dystrophy. The presence of CRALBP serves as a marker for RPE and Muller glial cells of the retina.
Specifications
| CRALBP | |
| Polyclonal | |
| Unconjugated | |
| RLBP1 | |
| 3110056M11Rik; cellular retinaldehyde-binding protein; cellular retinaldehyde-binding protein-1; CRALBP; retinaldehyde binding protein 1; retinaldehyde-binding protein 1; retinaldehyde-binding protein precursor; Rlbp1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 6017 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P12271 | |
| RLBP1 | |
| Recombinant protein corresponding to Human RLBP1. Recombinant protein control fragment (Product #RP-106021). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction