Learn More
Macherey-Nagel™ Crimping Head
Brand:  Macherey-Nagel™ 735720
 View more versions of this product
Product Code. 12898595
  • 457.00 EUR / Each
In Stock
Specifications
Crimping Head
1 Each
For Electronic High Power Crimping Tool 735700 and 20 mm Crimp Caps