Learn More
Fisherbrand™ Stir Bar Box Set, Cross-Shaped
Shop All Fisherbrand Products
Click to view available options
Length (Metric):
195 mm
Unit Size:
1 set
Specifications
Specifications
| Autoclavable | Autoclavable |
| Certifications/Compliance | Certificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS) |
| Color | White |
| Description | PTFE encapsulated Alnico V magnet core |
| Dimensions (L x W x H) | 195 x 95mm |
| For Use With (Equipment) | Containers & Vessels / Stirrer Hot Plates |
| Height (Metric) | 30mm |
| Includes | 5 Cross Stirrer Bars 1 each of 10x10, 20x20, 25x25, 30x30, 38x38 mm |
| Width (Metric) | 95mm |
| Length (Metric) | 195 mm |
| Show More |