Learn More
Invitrogen™ CTCF Monoclonal Antibody (G.758.4)
GREENER_CHOICE
Rabbit Monoclonal Antibody
Brand:  Invitrogen™ MA511187
Product Code. 10335688
  • 586.00 EUR / 100µL
Description
It is not recommended to aliquot this antibody. This antibody is not cross-reactive with BORIS.
Transcriptional factor CTCF is a ubiquitously expressed 11-zinc finger domain containing, DNA-binding nuclear phosphoprotein that is involved in multiple aspects of normal gene regulation including transcriptional repression and activation, gene silencing, chromatin insulation and regulation of imprinted sites. CTCF uses different combinations of ZF domains and interacts with the CCCTC motif on the DNA. It can bind to HAT- and HDAC-containing complexes thereby influencing transcriptional activation or repression respectively. It is a multifunctional protein involved in the transactivation of APPB gene, silencing of c-myc gene, and insulation of human beta-globin and DMI myotonic dystrophy locus and imprinting control of IGF2 and H19 regions. CTCF has also been shown to regulate the transcription of Pax6 and IRAK2 promoter thereby opening a therapeutic avenue. Mutations in this gene have been associated with invasive breast cancers, prostate cancers and Wilms' tumors.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CTCF
Monoclonal
91 μg/mL
0.01M HEPES with 0.15M NaCl, 100μg/mL BSA, 50% glycerol and <0.02% sodium azide; pH 7.5
P49711, Q61164, Q9R1D1
CTCF
Synthetic peptide corresponding to the carboxy terminus of the human CTCF protein.
100 μL
Primary
Human, Mouse, Monkey, Rat
Antibody
IgG
ChIP Assay, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
G.758.4
Unconjugated
CTCF
11 zinc finger transcriptional repressor; 11-zinc finger protein; AW108038; CCCTC-binding factor; CCCTC-binding factor (zinc finger protein); Ctcf; ctcf protein; CTCFL paralog; im:7152666; MRD21; OTTHUMP00000197610; Transcriptional repressor CTCF; Unknown (protein for IMAGE:8125283)
Rabbit
Affinity chromatography
RUO
10664, 13018, 83726
-20°C
Liquid
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions